missing translation for 'onlineSavingsMsg'
Learn More

LGD-2226, TRC

Product Code. 30011588
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011588 5 mg 5mg
30011352 10 mg 10mg
30003655 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011588 Supplier Toronto Research Chemicals Supplier No. TRCL3976155MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328947-93-9
Molecular Formula C14 H9 F9 N2 O
Molecular Weight (g/mol) 392.22
Synonym 6-[Bis(2,2,2-trifluoroethyl)amino]-4-(trifluoromethyl)-2(1H)-quinolinone
IUPAC Name 6-[bis(2,2,2-trifluoroethyl)amino]-4-(trifluoromethyl)-1H-quinolin-2-one
SMILES FC(F)(F)CN(CC(F)(F)F)c1ccc2NC(=O)C=C(c2c1)C(F)(F)F

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H9F9N2O/c15-12(16,17)5-25(6-13(18,19)20)7-1-2-10-8(3-7)9(14(21,22)23)4-11(26)24-10/h1-4H,5-6H2,(H,24,26)
Formula Weight 392.0571
Percent Purity >95%
Chemical Name or Material 6-[bis(2,2,2-trifluoroethyl)amino]-4-(trifluoromethyl)-1H-quinolin-2-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.