missing translation for 'onlineSavingsMsg'
Learn More

LGD-4033, TRC

Product Code. 30004477
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004477 25 mg 25mg
30005883 10 mg 10mg
30010352 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004477 Supplier Toronto Research Chemicals Supplier No. TRCL39762025MG
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1165910-22-4
Molecular Formula C14 H12 F6 N2 O
Molecular Weight (g/mol) 338.25
Synonym Benzonitrile, 4-[(2R)-2-[(1R)-2,2,2-trifluoro-1-hydroxyethyl]-1-pyrrolidinyl]-2-(trifluoromethyl)-, 4-[(2R)-2-[(1R)-2,2,2-Trifluoro-1-hydroxyethyl]-1-pyrrolidinyl]-2-(trifluoromethyl)benzonitrile, LGD-4033, VK5211, Ligandrol
IUPAC Name 4-[(2R)-2-[(1R)-2,2,2-trifluoro-1-hydroxyethyl]pyrrolidin-1-yl]-2-(trifluoromethyl)benzonitrile
SMILES O[C@H]([C@H]1CCCN1c2ccc(C#N)c(c2)C(F)(F)F)C(F)(F)F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H12F6N2O/c15-13(16,17)10-6-9(4-3-8(10)7-21)22-5-1-2-11(22)12(23)14(18,19)20/h3-4,6,11-12,23H,1-2,5H2/t11-,12-/m1/s1
Formula Weight 338.0854
Percent Purity >95%
Chemical Name or Material LGD-4033
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.