missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Lidocaine

Product Code. 18746426
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18746426 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18746426 Supplier Tocris Bioscience™ Supplier No. 3057/10
  • ONLINE EXCLUSIVE PRICE
    71.00 €
    67.20 € / 10mg
    Save 3.80 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anasthetic and class Ib antiarrhythmic agent. Blocks voltage-gated sodium channels in the inactivated state.

Lidocaine is an anasthetic and class Ib antiarrhythmic agent. Blocks voltage-gated sodium channels in the inactivated state.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Diethylamino-N-(2,6-dimethylphenyl)acetamide
CAS 137-58-6
Quantity 10 mg
Molecular Formula C14H22N2O
Formula Weight 234.34
Product Type Channel Blocker
Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.