missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine, MP Biomedicals

Product Code. 11484552 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484552 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11484552 Supplier MP Biomedicals Supplier No. 0219011125
  • ONLINE EXCLUSIVE PRICE
    66.45 €
    63.00 € / 25g
    Save 3.45 €
    5% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium channel blocker and class IB antiarrhythmic agent.

Specifications

CAS 137-58-6
Quantity 25 g
Molecular Formula C14H22N2O
Synonym lidocaine, lignocaine, xylocaine, lidoderm, 2-diethylamino-n-2,6-dimethylphenyl acetamide, anestacon, esracaine, duncaine, cappicaine, gravocain
InChI Key NNJVILVZKWQKPM-UHFFFAOYSA-N
SMILES CCN(CC)CC(=O)NC1=C(C=CC=C1C)C
IUPAC Name 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide
Molecular Weight (g/mol) 234.343
PubChem CID 3676
ChEBI CHEBI:6456
Grade Not Available
Chemical Name or Material Lidocaine
Show More Show Less
compliance-icons
Product Identifier
  • Lidocaine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Lidocaine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.