missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine, TRC

Product Code. 17971655
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971655 1 g 1g
17923465 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17971655 Supplier Toronto Research Chemicals Supplier No. TRCL3978051G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-58-6
Molecular Formula C14 H22 N2 O
Molecular Weight (g/mol) 234.34
Synonym Lidocaine, Acetamide, 2-(diethylamino)-N-(2,6-dimethylphenyl)-, 2',6'-Acetoxylidide, 2-(diethylamino)- (8CI), α-Diethylamino-2,6-acetoxylidide, 2-(Diethylamino)-N-(2,6-dimethylphenyl)acetamide, 2-(Diethylamino)-2',6'-acetoxylidide, Anestacon, Aritmal, Cuivasil, Dalcaine, Duncaine, ELA-Max, Esracaine, Isicaina, Isicaine, Jetocaine, LMX, Leostesin, Lida-Mantle, LidoPosterine, Lidocadren, Lidocain, Lidoderm, Lignocaine, Linisol, Maricaine, Medicaine, NSC 40030, Penles, Remicaine, Rucaina, Solarcaine, Solcain, Trachisan, Versatis, Xilina, Xybrex, Xycaine, Xylestesin, Xyline, Xylocain, Xylocaine, Xylocitin, Xylovet
IUPAC Name 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide
SMILES CCN(CC)CC(=O)Nc1c(C)cccc1C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H22N2O/c1-5-16(6-2)10-13(17)15-14-11(3)8-7-9-12(14)4/h7-9H,5-6,10H2,1-4H3,(H,15,17)
Formula Weight 234.1732
Percent Purity >95%
Chemical Name or Material Lidocaine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.