missing translation for 'onlineSavingsMsg'
Learn More

BOLA Lids

Product Code. 10296971
Change view
Click to view available options
Diameter (Metric) Outer:
60 mm
66 mm
82 mm
125 mm
166 mm
185 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer unitSize
10296971 60 mm Each
10104451 66 mm Each
10290092 82 mm Each
10076841 125 mm Each
10504022 166 mm Each
10737342 185 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10296971 Supplier BOLA Supplier No. H92707
  • ONLINE EXCLUSIVE PRICE
    25.20 €
    19.23 € / Pack of 1
    Save 5.97 €
    23% Off
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Lids PTFE, tight-closing

Lid with centering shoulder.

Specifications

Diameter (Metric) Outer 60 mm
Autoclavable Autoclavable
Disposable Reusable
Certifications/Compliance FDA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.