missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ Lids for Collection Pitchers

Product Code. 15213059
Change view
Click to view available options
No. per Pack:
6/Pk.
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Pack Unit Size
15213059 6/Pk. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15213059 Supplier Vitlab™ Supplier No. 97791
  • ONLINE EXCLUSIVE PRICE
    28.50 €
    20.60 € / Pack of 6
    Save 7.90 €
    27% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Constructed from Polypropylene. Vitlab™ Lids for Collection Pitchers are ideal for covering beakers used in chemical reactions.

  • For use with Polypropylene and SAN Collection Pitchers
  • White PC construction
TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Equipment) Collection pitcher
No. per Pack 6/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.