missing translation for 'onlineSavingsMsg'
Learn More

Liensinine, TRC

Product Code. 30484108
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30484108 2 mg 2mg
30482781 1 mg 1mg
30469816 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30484108 Supplier Toronto Research Chemicals Supplier No. TRCL3978332MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2586-96-1
Molecular Formula C37H42N2O6
Molecular Weight (g/mol) 610.74
Synonym 4-[[(1R)-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-1-isoquinolinyl]methyl]-2-[[(1R)-1,2,3,4-tetrahydro-1-[(4-hydroxyphenyl)methyl]-6-methoxy-2-methyl-7-isoquinolinyl]oxy]-Phenol, [R-(R*,R*)]-4-[(1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-1-isoquinolinyl)methyl]-2-[[1,2,3,4-tetrahydro-1-[(4-hydroxyphenyl)methyl]-6-methoxy-2-methyl-7-isoquinolinyl]oxy]-Phenol, 4-[[(1R)-1,2,3,4-Tetrahydro-6,7-dimethoxy-2-methyl-1-isoquinolinyl]methyl]-2-[[(1R)-1,2,3,4-tetrahydro-1-[(4-hydroxyphenyl)methyl]-6-methoxy-2-methyl-7-isoquinolinyl]oxy]phenol
SMILES COc1cc2CCN(C)[C@H](Cc3ccc(O)c(Oc4cc5[C@@H](Cc6ccc(O)cc6)N(C)CCc5cc4OC)c3)c2cc1OC

Specifications

Quantity 2 mg
InChI Formula InChI=1S/C37H42N2O6/c1-38-14-13-26-20-35(43-4)37(22-29(26)30(38)16-23-6-9-27(40)10-7-23)45-33-18-24(8-11-32(33)41)17-31-28-21-36(44-5)34(42-3)19-25(28)12-15-39(31)2/h6-11,18-22,30-31,40-41H,12-17H2,1-5H3/t30-,31-/m1/s1
Chemical Name or Material Liensinine
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.