missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIF Mouse anti-Human, Clone: 9824, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB250-SP
This item is not returnable.
View return policy
Description
LIF Monoclonal specifically detects LIF in Human samples. It is validated for Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| LIF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| LIF | |
| E. coli-derived recombinant human LIF | |
| 25 μg | |
| Primary | |
| Detects human LIF in ELISAs and Western blots. In sandwich immunoassays, no significant cross-reactivity or interference with recombinant human (rh) IL-1 alpha , rhIL-1 beta , rhIL-2, rhIL-3, rhIL-4, rhIL-6, rhIL-7, rhIL-8, rhG-CSF, rhGM-CSF, rhOSM, rhTGF-beta 1, rhTNF-alpha , rhTNF-beta , recombinant mouse (rm) IL-1 beta , rmIL-3, rmIL-4, rmIL-5, rmIL-6, rmIL-7, rmGM-CSF, bovine (b) FGF acidic, bFGF basic, human (h) PDGF, porcine (p) PDGF, hTGF-beta 1, pTGF-beta 1.2, or pTGF-beta 2 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry, Neutralization, ELISA | |
| 9824 | |
| Immunohistochemistry 8-25 ug/mL, Neutralization 0.06-0.2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3976 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction