missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Calibration Install Kit, Fast 96-well

Product Code. 11662119 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11662119 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11662119 Supplier Applied Biosystems™ Supplier No. 4432557

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Background plate; ROI calibration plate; Six spectral calibration plates with FAM™, SYBR™ Green, VIC™, NED™, TAMRA™, and ROX™ dye standards; Two normalization plates; RNase P instrument verification plate

This kit is used to maintain your ViiA™ 7 System with Fast 96-well Block.

It includes calibration plates to perform a spectral calibration with FAM™, VIC™, ROX™, SYBR™ Green, TAMRA™, NED™ dyes, plates to perform region-of-interest (ROI) calibration, normalization calibration, and to run an RNase P verification. Calibration and verification should be run at least every six months. Please see your instrument maintenance guide for full instructions on use. The dye formulations for your ViiA™ 7 System will improve results with multiplexing by more accurately representing fluorescent spectra found in your real-time PCR experiments.

Calibration and verification should be done at least every six months. Please see instrument maintenance guide for full instructions.

Order Info

Store kit at -15° to -25°C

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Includes:
• 1 background calibration plate
• 1 ROI calibration plate
• 6 spectral calibration plates with FAM, SYBR Green, VIC, NED, TAMRA, and ROX dye standards
• 2 normalization calibration plates
• 1 RNase P instrument verification plate

Store kit at -5 to -30°C.
For Use With (Equipment) QuantStudio™ 12 K Flex, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System
Product Type Calibration Install Kit
For Use With (Application) Real Time PCR (qPCR)
Quantity 1 kit
Calibration Type Spectral Calibration

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.