Learn More
Invitrogen™ E-Gel™ Imager System with UV Light Base
Description
Includes
1 x E-Gel Imager Camera Hood, 1 x E-Gel Imager UV Light Base, 1 x E-Gel Imager software package, 1 x Gel Quant Express software enabling USB dongle, 1 x E-Gel iBase light diffuser
Requires
Minimum System Requirements for GelCapture and Gel Quant Express: Processor: Intel™ Core™2 Duo 1.8GHz; Memory: 1Gb Ram; OS: Windows™ XP Professional SP3 (English version) or Windows™ 7 Professional (English version), must have administrator permissions for installation. Monitor: 24 x 786 minimum resolution; USB: USB 2.0 port for software dongle
Each E-Gel Imager system includes a sleek and compact camera hood and one or more interchangeable bases along with two powerful software programs.
- Captures sharp, rich images with the integrated scientific grade camera
- Captures gel images in real time, even during a run
- Analyzes gel images using the powerful Gel Quant Express software
- Powerful and Compact Scientific Grade Camera
- E-Gel Imager Camera Hood is powerhouse at core of E-Gel Imager system featuring a high resolution, scientific grade camera
- Captures robust gel images with maximum dynamic range and sensitivity
- UV Light Base is best for ethidium bromide and the Blue Light Base can be used for many SYBR™ dyes like SYBR Safe dye
- Gel Compatibility: Agarose Gels, E-Gel™, E-Gel™ 96; Label or Dye: Qdot™ 625, SYBR™
- Two powerful software tools included with the E-Gel Imager system, GelCapture and Gel Quant Express
- GelCapture allows to adjust sensitivity and exposure settings to capture the perfect image of your gel
- Perfect image basic manipulations like inversion or pixel coloration can be performed
- Gel Quant Express software allows quantitation and analysis via 1D gel analysis or using the express mode
- GelCapture can be downloaded and utilized on any PC, while Gel Quant Express requires a USB dongle for operation
- Additional USB dongles can be purchased to use Gel Quant Express on more than one PC simultaneously
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
Specifications
| Content And Storage | 1 x E-Gel™ Imager Camera Hood. 1 x E-Gel™ Imager UV Light Base. 1 x E-Gel™ Imager software package. 1 x Gel Quant Express software enabling USB dongle. 1 x E-Gel Imager light diffuser. 1 x E-Gel Imager universal filter. Store at room temperature. |
| For Use With (Equipment) | E-Gel Imager |
| Gel Compatibility | E-Gel™, E-Gel™ 48, E-Gel™ 96, Agarose Gels |
| Product Line | E-Gel |
| Type | Imager System with UV Light Base |
| Label or Dye | Ethidium bromide |
| System Type | E-Gel™ Imager |
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.