Learn More
Cytiva LifescienceManual Injection Valve
Product Code. 15239774 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
AKTA start Automated Protein Purification System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15239774 AKTA start Automated Protein Purification System Each
1 options
Product Code. 15239774 Supplier Cytiva Supplier No. 29023958
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with the AKTA start automated protein purification system. The manual injection valve is a 6-port rotary valve that is manually operated to transfer the preloaded sample onto the column. The sample loop (for the injection of small sample volumes [25μL to 5mL], or a Superloop injection loop for larger sample volumes [10 to 150mL]), can easily be connected to the appropriate ports of the valve.
TRUSTED_SUSTAINABILITY
Specifications
Description 6-port rotary valve that is manually operated to transfer the preloaded sample onto the column
For Use With (Equipment) AKTA start Automated Protein Purification System