missing translation for 'onlineSavingsMsg'
Learn More

Purolite™ Lifetech™ ECR Resins for Enzyme Immobilization, 150 to 300 μm

Product Code. 17099373
Change view
Click to view available options
Quantity:
250 g
1 kg
Pore Size:
1200 to 1800 Å
300 to 600 Å
600 to 1200 Å
Unit Size:
1kg
250g
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Pore Size unitSize
17099373 1 kg 1200 to 1800 Å 1kg
17009333 250 g 600 to 1200 Å 250g
17009363 250 g 1200 to 1800 Å 250g
17019333 1 kg 600 to 1200 Å 1kg
17019353 250 g 600 to 1200 Å 250g
17019363 1 kg 1200 to 1800 Å 1kg
17029353 1 kg 600 to 1200 Å 1kg
17029373 250 g 600 to 1200 Å 250g
17039373 1 kg 600 to 1200 Å 1kg
17049323 250 g 300 to 600 Å 250g
17059323 1 kg 300 to 600 Å 1kg
17059343 250 g 300 to 600 Å 250g
17069333 250 g 1200 to 1800 Å 250g
17069343 1 kg 300 to 600 Å 1kg
17069363 250 g 300 to 600 Å 250g
17079333 1 kg 1200 to 1800 Å 1kg
17079363 1 kg 300 to 600 Å 1kg
17089373 250 g 1200 to 1800 Å 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Product Code. 17099373 Supplier Purolite™ Supplier No. LS0139194

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lifetech™ ECR resins are robust polymeric resins designed for research and industrial immobilization of enzymes. Available for covalent, ionic, adsorption and affinity immobilization.

  • Enzyme immobilization by adsorption, covalent, IEX and affinity
  • High mechanical stability
  • High porosity
  • Easily dried after immobilization
  • Suitable for batch and column configuration
  • Optimal for use in organic solvent/water free systems
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free

Specifications

pH Range 3 to 10
Pore Size 1200 to 1800 Å
Content And Storage 2°C to 20°C
Particle Size 150 to 300 μm
Moisture 70 to 80%
Functionality/Interaction Type Amino
Form Wet
Product Line Lifetech
Quantity 1 kg
Type Methacrylate (ECR Resin)
Format White to Off-White Spherical Beads
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.