missing translation for 'onlineSavingsMsg'
Learn More

Purolite™ Lifetech™ ECR Resins for Enzyme Immobilization, 300 to 710 μm

Product Code. 17009343
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Pore Size:
1200 to 1800 Å
220 to 340 Å
300 to 600 Å
400 to 650 Å
600 to 1200 Å
Unit Size:
1kg
250g
50g
27 product options available for selection
Product selection table with 27 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Pore Size unitSize
17009343 1 kg 1200 to 1800 Å 1kg
17009373 1 kg 300 to 600 Å 1kg
17019323 250 g 220 to 340 Å 250g
17019383 250 g 1200 to 1800 Å 250g
17029323 1 kg 220 to 340 Å 1kg
17029383 1 kg 1200 to 1800 Å 1kg
17039333 250 g 600 to 1200 Å 250g
17039363 250 g 1200 to 1800 Å 250g
17049333 1 kg 600 to 1200 Å 1kg
17049353 250 g 600 to 1200 Å 250g
17049363 1 kg 1200 to 1800 Å 1kg
17049383 250 g 400 to 650 Å 250g
17059353 1 kg 600 to 1200 Å 1kg
17059373 250 g 600 to 1200 Å 250g
17059383 1 kg 400 to 650 Å 1kg
17069373 1 kg 600 to 1200 Å 1kg
17079313 50 g - 50g
17079323 250 g 300 to 600 Å 250g
17079353 250 g 600 to 1200 Å 250g
17089313 250 g - 250g
17089323 1 kg 300 to 600 Å 1kg
17089343 250 g 300 to 600 Å 250g
17089353 1 kg 600 to 1200 Å 1kg
17099313 1 kg - 1kg
17099333 250 g 1200 to 1800 Å 250g
17099343 1 kg 300 to 600 Å 1kg
17099363 250 g 300 to 600 Å 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
27 options
This item is not returnable. View return policy
Product Code. 17009343 Supplier Purolite™ Supplier No. LS0124594

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lifetech™ ECR resins are robust polymeric resins designed for research and industrial immobilization of enzymes. Available for covalent, ionic, adsorption and affinity immobilization.

  • Enzyme immobilization by adsorption, covalent, IEX and affinity
  • High mechanical stability
  • High porosity
  • Easily dried after immobilization
  • Suitable for batch and column configuration
  • Optimal for use in organic solvent/water free systems
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free

Specifications

pH Range 5 to 9
Pore Size 1200 to 1800 Å
Content And Storage 2°C to 8°C
Particle Size 300 to 710 μm
Moisture 70 to 80%
Functionality/Interaction Type Epoxy
Form Wet
Product Line Lifetech
Quantity 1 kg
Type Methacrylate (ECR Resin)
Format White to Off-White Spherical Beads
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.