missing translation for 'onlineSavingsMsg'
Learn More

Purolite™ Lifetech™ ECR Resins for Enzyme Immobilization, ECR8285

Product Code. 17039343
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17039343 1 kg 1kg
17029343 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17039343 Supplier Purolite™ Supplier No. LS0126994

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lifetech™ ECR resins are robust polymeric resins designed for research and industrial immobilization of enzymes. Available for covalent, ionic, adsorption and affinity immobilization.

  • Enzyme immobilization by adsorption, covalent, IEX and affinity
  • High mechanical stability
  • High porosity
  • Easily dried after immobilization
  • Suitable for batch and column configuration
  • Optimal for use in organic solvent/water free systems
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free

Specifications

pH Range 5 to 9
Pore Size 400 to 600 Å
Content And Storage 2°C to 8°C
Particle Size 250 to 1000 μm
Moisture 70 to 80%
Functionality/Interaction Type Epoxy
Form Wet
Quantity 1 kg
Type Methacrylate (ECR Resin)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.