Learn More
Lift Pump with Downtube
Product Code. 12907974
Encompass
Click to view available options
Material:
Nylon
Polypropylene
Unit Size:
Each
Product Code. 12907974
This item requires a price update prior to being ordered.
Request a Price for this item here.
For 20L/25L containers, capacity 0.33L per stroke. For 205L containers capacity 0.5L per stroke. Available in two sizes. With a 900mm flexible dispensing tube, except polypropylene 205L dispensing pump.
  • Manufactured with nylon for maximum resistance to acids and solvents
  • No metal part comes into contact with the liquid being pumped
  • PTFE seals fitted
  • Universal adapters fitted to provide secure seating in the necks of virtually all drums from 20L/25L to 205L capacities
  • Piston action ensures that the liquid flow is totally under control at all times during the action For dispensing solvents and acids in controlled amounts.
Specifications
Quantity 1 Each
Type Lift Pump
For Use With (Application) For dispensing solvents and acids in controlled amounts
Material Nylon