missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Light Emission Accelerator-II

Product Code. 12694335
Change view
Click to view available options
Quantity:
210 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12694335 210 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12694335 Supplier Invitrogen™ Supplier No. T2222
  • ONLINE EXCLUSIVE PRICE
    376.00 €
    304.56 € / 210mL
    Save 71.44 €
    19% Off
View all promotions
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Light emission triggering solution with luminescence enhancer for use with the Galacto-Light Plus™ or Dual-Light™ Reporter Gene Assay Systems.

Light emission triggering solution with luminescence enhancer for use with the Galacto-Light Plus™ or Dual-Light™ Reporter Gene Assay Systems.

For Research Use Only. Not for use in diagnostics procedures.

  • Form: Liquid
  • Format: Bottle
  • Substrate: Alkaline Phosphatase
TRUSTED_SUSTAINABILITY

Specifications

Product Type β-Galactosidase Reporter Gene Assay System
Content And Storage Ambient for shipping. Store the product at 4°C.
Shipping Condition Room Temperature
Substrate Alkaline Phosphatase
Substrate Type AP (Alkaline Phosphatase) Substrate
Product Line NovaBright
Quantity 210 mL
Form Liquid
compliance-icons compliance-icons
Product Identifier
  • ACCELERATOR-II 210 ML
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Diethanolamine

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.