Learn More
Hirschmann™ Light Protection Window
Product Code. 11652239
Change view
Click to view available options
For Use With (Equipment):
opus™
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
11652239 opus™ Each
1 options
Product Code. 11652239 Supplier Hirschmann™ Supplier No. 9586060
  • ONLINE EXCLUSIVE PRICE
    24.26 €
    15.78 € / Pack of 1
    Save 8.48 €
    34% Off
In Stock
Add to basket
Add to basket
Amber colored protection window for UV-sensitive media, used with opus™ and solarus™.
  • Amber colored replacement observation window
  • For the protection of UV-sensitive media
Specifications
Description For light sensitive media
Type UV Protection Window
Includes 1
For Use With (Equipment) opus™