Learn More
Thermo Scientific™ Light /UV Protection for Vertical Light Chambers
Product Code. 16461125 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
Factory Installed Door Switch
Unit Size:
Each
Product Code. Includes Unit Size
16461125 Factory Installed Door Switch Each
1 options
Product Code. 16461125 Supplier Thermo Scientific™ Supplier No. 1900663
  • ONLINE EXCLUSIVE PRICE
    119.92 €
    108.00 € / Pack of 1
    Save 11.92 €
    9% Off
 Restricted Item
Estimated Shipment: 03-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Protection for operator, light switches off automatically when door is opened
Light / UV protection is a feature to protect the operator's eyes. This factory-installed option will switch off the vertical light as the door is opened. Recommended for ICH photostability testing. Not recommended for plant growth / animal hatching, sinc
  • Light / UV protection for operator's eyes.
  • Factory-installed option will switch off the vertical light as the door is opened
  • Recommended for ICH photo stability testing
  • Not recommended for plant growth/animal hatching since switching light on and off could stress plants and animals
  • Operator can wear goggles as an alternative protection.
TRUSTED_SUSTAINABILITY
Specifications
Description Light / UV protection - light off with open door
For Use With (Equipment) All Thermo Scientific and Forma vertical light chambers - models 3906, 3909, 3928, 3942, 3943
Lighting LED
Type Light/UV Protection
For Use With (Application) Light application - recommended for ICH photostability testing
Includes Factory Installed Door Switch