missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies LightCompass™ Plus Data Acquisition, Analysis and Reporting Software

Product Code. 31777369 Shop All Berthold Technologies Products
Change view
Click to view available options
Product Type:
Software for Microplate Reader
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
31777369 Software for Microplate Reader Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31777369 Supplier Berthold Technologies Supplier No. 37854404
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Scientific software for data acquisition, analysis, and reporting with flexible protocols, curve fitting, qualitative analysis, and automation.

Workflow

Protocol setup

  • Create or open a plate layout.
  • Open a preloaded protocol (includes instrument settings, analysis functions, and export/print settings), use it as a template, or build a new one with the protocol wizard.
  • Create fully custom protocols if detailed control is required.

Measurement capabilities

  • Reagent dispensing and incubation steps
  • Endpoint
  • Kinetics (including long-term/repeated measures over several days)
  • Wavelength scan
  • Area scan
  • Multiple measurement modes within a single protocol

Data analysis

  • Standard curves with curve-fitting options: linear, polynomial, 4PL, smoothed cubic spline, others
  • Threshold-based classification for qualitative assays
  • Custom formulas
  • Quantitative and qualitative analysis tools
  • Outlier correction
  • Unlimited calculation matrices
  • Dilution factor calculation
  • Customized analysis report generation

Data handling and compliance

  • Print or export results to Excel or LIMS
  • User accounts with configurable levels, hierarchies, and password protection
  • Active Directory integration
  • Audit trail with automated log files
  • QC/validation settings with pass/fail criteria
  • Secure data storage (local file system or remote SQL server)
  • FDA 21 CFR Part 11 and EU GMP Annex 11 compliance
  • Control history for validation
  • Assay lot number verification

Sample and plate management

  • Sample identification (manual or barcode)
  • Plate Organizer (sample ID definition, preparation)
  • Automated filename generation
  • Parameter file selection by barcode
  • Automation Server support

Applications

  • ELISA
  • Nucleic acid and protein quantification
  • Reporter gene assays
  • Dose-response assays
  • Kinetic assays
  • BRET
  • TR-FRET and HTRF™

Specifications

Product Type Software for Microplate Reader
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.