missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ tRNA for LightShift™ Chemiluminescent RNA EMSA Kit

Product Code. 10629389 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10629389 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10629389 Supplier Thermo Scientific™ Supplier No. 20159
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Study RNA-protein interactions with this non-radioactive EMSA kit for gel shift assays.

Thermo Scientific™ LightShift Chemiluminescent RNA EMSA Kit provides a non-radioactive solution for studying RNA-protein interactions using an electrophoretic mobility-shift assay (EMSA).

The RNA EMSA Kit uses biotinylated RNA probes and a chemiluminescent substrate system to achieve fast and safe detection of RNA-protein complexes with sensitivity equivalent to traditional 32P-isotopic methods. The complete kit comes with all reagents required to set up and optimize protein-RNA binding conditions, a positive control for protein-RNA interactions and reagents for chemiluminescent detection of the nucleic acid interaction.

Highlights:

  • Sensitive – chemiluminescent detection comparable to radioactive detection

  • Time-saving – develop X-ray films after 1- to 5-minute exposures, versus 16 hour exposures needed with radioactive systems

  • Flexible – compatible with RNA probes biotinylated by different methods

  • Easy to use – complete kit includes optimized reagents for binding reactions and RNA probe detection

  • Non-radioactive – eliminate waste concerns from radioactive RNA probes

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -70°C.
Detection Method Chemiluminescence
Format Kit
Product Line LightShift
Product Type tRNA for LightShift RNA EMSA Kit
Quantity 100 μL
Target Specificity Not Target-Specific

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.