missing translation for 'onlineSavingsMsg'
Learn More

Ligustrazine, MedChemExpress

Product Code. 16469036
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469036 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16469036 Supplier Medchem Express Supplier No. HYN0264
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ligustrazine (Chuanxiongzine), an alkylpyrazine isolated from Ligusticum wallichii (Chuan Xiong), is present in french fries, bread, cooked meats, tea, cocoa, coffee, beer, spirits, peanuts, filberts, dairy products and soy products as fragrance and flavouring ingredienexhibits. Ligustrazine also has potential nootropic and anti-inflammatory activities in rats.

Ligustrazine (Chuanxiongzine), an alkylpyrazine isolated from Ligusticum wallichii (Chuan Xiong), is present in french fries, bread, cooked meats, tea, cocoa, coffee, beer, spirits, peanuts, filberts, dairy products and soy products as fragrance and flavouring ingredienexhibits. Ligustrazine also has potential nootropic and anti-inflammatory activities in rats.

Specifications

Chemical Name or Material Ligustrazine
CAS 1124-11-4
Color White
Purity Grade Notes Research
Molecular Formula C8H12N2
Quantity 100 mg
Synonym Chuanxiongzine Tetramethylpyrazine
Solubility Information DMSO : 50 mg/mL (367.13 mM; Need ultrasonic)
SMILES CC1=C(C)N=C(C)C(C)=N1
Molecular Weight (g/mol) 136.19
Formula Weight 136.19
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.