missing translation for 'onlineSavingsMsg'
Learn More

Limaprost, MedChemExpress

Product Code. 16481038
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481038 5 mg 5mg
16454447 50 mg 50mg
16440376 2 mg 2mg
16428578 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481038 Supplier Medchem Express Supplier No. HYB0683
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Limaprost (OP1206) is a PGE1 analogue and a potent and orally active vasodilator. Limaprost increases blood flow and inhibits platelet aggregation. Limaprost pain relief, has antianginal effects, and can be used for ischaemic symptoms research.

Limaprost (OP1206) is a PGE1 analogue and a potent and orally active vasodilator. Limaprost increases blood flow and inhibits platelet aggregation. Limaprost pain relief, has antianginal effects, and can be used for ischaemic symptoms research.

Specifications

Chemical Name or Material Limaprost
CAS 74397-12-9
Color White
Purity Grade Notes Research
Molecular Formula C22H36O5
Quantity 5 mg
Synonym 17α,20-dimethyl-δ2-PGE1 ONO1206 OP1206
Solubility Information DMSO : ≥ 40 mg/mL (105.12 mM)
SMILES O=C(O)/C=C/CCCC[C@@H]1[C@@H](/C=C/[C@@H](O)C[C@@H](C)CCCC)[C@H](O)CC1=O
Molecular Weight (g/mol) 380.52
Formula Weight 380.52
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.