missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LIMKi 3

Description
LIMKi 3 is a potent LIM kinase inhibitor (IC50 values are 7 and 8 nM for LIMK1 and LIMK2 respectively). Inhibits cofilin phosphorylation in MDA-MB-231 breast cancer cells. Reduces MDA-MB-231 tumor cell invasion in a 3D matrigel invasion assay.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | LIMKi 3 |
| Purity | >98% |
| Molecular Weight (g/mol) | 431.29 |
| Inhibitors | LIMKi 3 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?