missing translation for 'onlineSavingsMsg'
Learn More

(+)-Limonene 2,3,3,5,5,-d5 (Major), TRC

Product Code. 30467329
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30467329 5 mg 5mg
30465393 1 mg 1mg
30466811 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30467329 Supplier Toronto Research Chemicals Supplier No. TRCL4617435MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2514937-17-6
Molecular Formula C10H11D5
Molecular Weight (g/mol) 141.26
Synonym (R)-Cyclohexene 1-methyl-4-(1-methylethenyl)-d5, (R)-(+)-p-Mentha-1,8-diene-d5 (8CI), (4R)-1-Methyl-4-(1-methylethenyl)cyclohexene-d5, (+)-(4R)-Limonene-d5, (+)-(R)-Limonene-d5, (+)-Dipentene-d5, (+)-Limonene-d5, (+)-p-Mentha-1,8-diene-d5, (+)-a-Limonene-d5, (4R)-(+)-Limonene-d5, (4R)-Limonene-d5, (R)-(+)-Limonene-d5, (R)-(+)-p-Mentha-1,8-diene-d5, (R)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-1-ene-d5, (R)-4-Isopropenyl-1-methyl-1-cyclohexene-d5, (R)-Limonene-d5, (R)-p-Mentha-1,8-diene-d5, 1-Methyl-4-prop-1-en-2-yl-cyclohexene-d5, Bio Clear-d5, Biogenic SE 374-d5, Carvene-d5, Clearene-d5, D-(+)-Limonene-d5, D-Limonene-d5, EC 7-d5, Fruits Dry-d5, Glidesafe-d5, Glidsafe-d5, Histo-Clear-d5, HistoSolve X-d5, IVOD-d5, Master Clear-d5, NK 1100-d5, Refchole-d5
IUPAC Name (R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene-2,3,3,5,5-d5
SMILES CC([C@@H]1C([2H])([2H])CC(C)=C([2H])C1([2H])[2H])=C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,10H,1,5-7H2,2-3H3/t10-/m0/s1/i4D,6D2,7D2
Formula Weight 141.16
Percent Purity >95%
Chemical Name or Material (+)-Limonene 2,3,3,5,5,-d5
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.