missing translation for 'onlineSavingsMsg'
Learn More

(+)-Limonene, TRC

Product Code. 30467015
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467015 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467015 Supplier Toronto Research Chemicals Supplier No. TRCL461740100ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (+)-Limonene
CAS 5989-27-5
Purity Grade Notes GC
Molecular Formula C10 H16
Quantity 100 mL
Synonym (4R)-4-Isopropenyl-1-methyl-cyclohexene, Cyclohexene, 1-methyl-4-(1-methylethenyl)-, (R)-, p-Mentha-1,8-diene, (R)-(+)- (8CI), (+)-(4R)-Limonene, (+)-(R)-Limonene, (+)-Dipentene, (+)-Limonene, (+)-p-Mentha-1,8-diene, (+)-α-Limonene, (4R)-(+)-Limonene, (4R)-1-Methyl-4-(1-methylethenyl)cyclohexene, (4R)-Limonene, (R)-(+)-Limonene, (R)-(+)-p-Mentha-1,8-diene, (R)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-1-ene, (R)-4-Isopropenyl-1-methyl-1-cyclohexene, (R)-Limonene, (R)-p-Mentha-1,8-diene, 1-Methyl-4-prop-1-en-2-yl-cyclohexene, Bio Clear, Biogenic SE 374, Carvene, Clearene, D-(+)-Limonene, D-Limonene, EC 7, Fruits Dry, Glidesafe, Glidsafe, Histo-Clear, HistoSolve X, IVOD, Limonene, (+)-, Master Clear, NK 1100, Refchole, Safsolv, Woody River 8, d-(+)-Limonene, d-Limonene
InChI Formula InChI=1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,10H,1,5-7H2,2-3H3/t10-/m0/s1
SMILES CC(=C)[C@@H]1CCC(=CC1)C
IUPAC Name (4R)-1-methyl-4-prop-1-en-2-ylcyclohexene
Molecular Weight (g/mol) 136.23
Formula Weight 136.1252
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.