missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Limonin
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Alpha Vector | LIMONIN |
| Melting Point | 298°C to 300°C |
| Color | White |
| Loss on Drying | ≤5 % |
| Infrared Spectrum | Conforms |
| Packaging | glass bottle |
| Molecular Formula | C26 H30 O8 |
| Quantity | 1 g |
| Formula Weight | 470.52 |
| Physical Form | Powder |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur