missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LIN28 1632, >98%

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Lin28 1632 is an RNA binding protein Lin28 inhibitor (IC50 = 8 μM). Lin28 1632 blocks binding of Lin28 to let-7 pre-micro RNA. Lin28 1632 inhibits stemness and induces differentiation of murine ESCs. Lin28 1632 increases let-7 miRNA and suppresses PD-L1 expression in multiple human cancer cell lines. Lin28 1632 inhibits growth of tumor cell lines and tumor-sphere formation in vitro. Lin28 1632 also inhibits growth of tumor xenografts in mice by enhancing T cell-mediated antitumor immunity . Lin28 1632 is also bromodomain inhibitor. Lin28 1632 inhibits tumor growth and metastasis in vivo in immunodeficient orthotopic mouse models of human breast cancer cell MDA-MB-231.
Specifications
Specifications
| Chemical Name or Material | N-Methyl-N-[3-(3-methyl-1,2,4-triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide |
| CAS | 108825-65-6 |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Molecular Formula | C15H15N5O |
| Formula Weight | 281.31 |
| Purity | >98% |
| Molecular Weight (g/mol) | 281.319 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction