missing translation for 'onlineSavingsMsg'
Learn More

Linalool, 97%

Product Code. 10387050 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10387050 100 mL Glass bottle 100mL
10746151 500 mL Glass bottle 500mL
10464975 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10387050 Proveedor Thermo Scientific Acros N.º de proveedor 125151000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 78-70-6 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 78-70-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00008906
InChI Key CDOSHBSSFJOMGT-UHFFFAOYSA-N
Synonym linalool, linalol, 3,7-dimethyl-1,6-octadien-3-ol, allo-ocimenol, beta-linalool, linalyl alcohol, 1,6-octadien-3-ol, 3,7-dimethyl, +--linalool, p-linalool, linanool
PubChem CID 6549
ChEBI CHEBI:17580
IUPAC Name 3,7-dimethylocta-1,6-dien-3-ol
SMILES CC(=CCCC(C)(C=C)O)C

Especificaciones

Color Colorless to Yellow
Density 0.8610g/mL
Boiling Point 199.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=CHCH2CH2C(CH3)(OH)CH=CH2
Refractive Index 1.4600 to 1.4640
Quantity 100 mL
Beilstein 01, 462
Merck Index 15, 5550
Specific Gravity 0.861
Specific Rotation -2° to +2° (20°C, 589nm) (neat)
Solubility Information Solubility in water: 1.45g/L (25°C). Other solubilities: miscible with alcohol and ether
Viscosity 4.4 mPa.s (25°C)
Formula Weight 154.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Linalool, 97%
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Linalool
Palabra de advertencia
  • Warning
Clasificación
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.