missing translation for 'onlineSavingsMsg'
Learn More

Linalool, TRC

Product Code. 17974815
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17974815 100 g 100g
17913985 10 g 10g
17962965 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17974815 Supplier Toronto Research Chemicals Supplier No. TRCL465950100G
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-70-6
Molecular Formula C10 H18 O
Molecular Weight (g/mol) 154.249
Synonym 1,6-Octadien-3-ol, 3,7-dimethyl-, (±)-Linalool, 2,6-Dimethyl-2,7-octadien-6-ol, 2-Methyl-1-prenyl-3-buten-2-ol, 3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethyl-1,6-octadiene-3-ol, 3,7-Dimethyl-3-hydroxy-1,6-octadiene, 3,7-Dimethyl-3-ol-1,6-octadiene, L 260-2, Linalol, Linalool, Linalyl alcohol, Linanool, NSC 3789, Phantol, dl-Linalool, β-Linalool
IUPAC Name 3,7-dimethylocta-1,6-dien-3-ol
SMILES CC(=CCCC(C)(O)C=C)C

Specifications

Quantity 100 g
InChI Formula InChI=1S/C10H18O/c1-5-10(4,11)8-6-7-9(2)3/h5,7,11H,1,6,8H2,2-4H3
Formula Weight 154.136
Chemical Name or Material DL-Linalool
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.