missing translation for 'onlineSavingsMsg'
Learn More

Linalyl Acetate, TRC

Product Code. 30132510
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132510 5 g 5g
30010360 500 mg 500mg
30007112 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132510 Supplier Toronto Research Chemicals Supplier No. TRCL2470455G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-95-7
Molecular Formula C12 H20 O2
Molecular Weight (g/mol) 196.29
Synonym 1,6-Octadien-3-ol, 3,7-dimethyl-, acetate (8CI,9CI), Linalool acetate (6CI), (±)-Linaloyl acetate, (±)-Linalyl acetate, 1,5-Dimethyl-1-vinyl-4-hexenyl acetate, 3,7-Dimethyl-1,6-octadien-3-yl acetate, 3-Acetoxy-3,7-dimethyl-1,6-octadiene, Acetic acid linalool ester, Bergamiol, Bergamol, Bergamot mint oil, Linalyl acetate, NSC 2138, dl-Linalool acetate
IUPAC Name 3,7-dimethylocta-1,6-dien-3-yl acetate
SMILES CC(=CCCC(C)(OC(=O)C)C=C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H20O2/c1-6-12(5,14-11(4)13)9-7-8-10(2)3/h6,8H,1,7,9H2,2-5H3
Formula Weight 196.1463
Chemical Name or Material Linalyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.