missing translation for 'onlineSavingsMsg'
Learn More

Linaprazan, MedChemExpress

Product Code. 16499566
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499566 50 mg 50mg
16405747 5 mg 5mg
16404316 100 mg 100mg
16403008 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499566 Supplier Medchem Express Supplier No. HY100412
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Linaprazan (AZD0865) inhibits gastric H+,K+-ATPase by K+-competitive binding.

Linaprazan (AZD0865) inhibits gastric H+,K+-ATPase by K+-competitive binding.

Specifications

Chemical Name or Material Linaprazan
CAS 248919-64-4
Color White
Purity Grade Notes Research
Molecular Formula C21H26N4O2
Quantity 50 mg
Synonym AZD0865
Solubility Information DMSO : ≥ 35 mg/mL (95.51 mM)
SMILES O=C(C1=CN2C(C(NCC3=C(C)C=CC=C3C)=C1)=NC(C)=C2C)NCCO
Molecular Weight (g/mol) 366.46
Formula Weight 366.46
Percent Purity 98.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.