missing translation for 'onlineSavingsMsg'
Learn More

Lincomycin Sulfoxide, TRC

Product Code. 30135122
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135122 25 mg 25mg
30005837 2.5 mg 2.5mg
30008714 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135122 Supplier Toronto Research Chemicals Supplier No. TRCL46610025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22083-92-7
Molecular Formula C18H34N2O7S
Molecular Weight (g/mol) 422.54
Synonym (2S,4R)-N-((1R,2R)-2-Hydroxy-1-((2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylsulfinyl)tetrahydro-2H-pyran-2-yl)propyl)-1-methyl-4-propylpyrrolidine-2-carboxamide;1,6,8-Trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-(methylsulfinyl)-D-erythro-alpha-D-galacto-octopyranose;1,6,8-Trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-(methylsulfinyl)-D-erythro-alpha-D-galacto-octopyranose;[2S-(2alpha,4beta)]-1,6,8-Trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-1,6,8-trideoxy-6-[[(1-methyl-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-(methylsulfinyl)-D-erythro-alpha-D-galacto-octopyranose, trans- alpha- 1,6,8-trideoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-(methylsulfinyl)-D-erythro-D-galacto-octopyranose;Lincomycin S-oxide;
SMILES CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]2O[C@@H]([C@H](O)[C@@H](O)[C@H]2O)S(=O)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H34N2O7S/c1-5-6-10-7-11(20(3)8-10)17(25)19-12(9(2)21)16-14(23)13(22)15(24)18(27-16)28(4)26/h9-16,18,21-24H,5-8H2,1-4H3,(H,19,25)/t9-,10-,11+,12-,13+,14-,15-,16-,18-,28?/m1/s1
Chemical Name or Material Lincomycin Sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.