missing translation for 'onlineSavingsMsg'
Learn More

Lindane, TRC

Product Code. 30477636
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477636 50 g 50g
30476956 10 g 10g
30468692 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477636 Supplier Toronto Research Chemicals Supplier No. TRCL46599050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-89-9
Molecular Formula C6 H6 Cl6
Molecular Weight (g/mol) 290.83
Synonym (1α,2α,3β,4α,5α,6β)-1,2,3,4,5,6-Hexachlorocyclohexane, 1,2,3,4,5,6-Hexachlorocyclohexane, 666, Aalindan, Aficide, Agrocide, Agrocide III, Agrocide WP, Ameisenmittel Merck, Aparasin, Aphtiria, Aplidal, Arbitex, Arcotal S, BBH, BHC, BHC (insecticide), Ben-Hex, Benhexol, Bentox 10, Benzene hexachloride, Bercema-Spritz-Lindan 50, Bexol, Celanex, Chloresene, Codechine, DBH, Detmol Extract, Devoran, Dol Granule, Drilltox-Spezial Aglukon, ENT 7,796, Entomoxan, Esoderm, Fenoform forte, Forst-Nexen, Gamacid, Gamacide, Gamacide 20, Gamene, Gamma benzene hexachloride, Gamma-HCH, Gammalin, Gammalin 20, Gammaterr, Gammaxene, Gammexane, Gamoline, Geobilan, Geolin G 3, Gexane, HCC, HCCH, HCH, HGI, Heclotox, Hexa, Hexachloran, Hexachlorane, Hexachlorocyclohexane, Hexatin, Hexaverm, Hexicide, Hexyclan, Hilbeech, Hortex, Hungaria L 7, Inexit, Jacutin, Kanodane, Kokotine, Kwell, Lasochron, Lendine, Lentox, Lidenal, Lindafor, Lindane, Lindatox, Lindex, Lindosep, Lintox, Lintox 3, Linvur, Lorexane, Mglawik L, Milbol 49, Mszycol, Neo-Scabicidol, Nexen FB, Nexit, Nexit Stark, Nexol E, Nicochloran, Omnitox, Ovadziak, Owadziak, PLK, Pedraczak, Pflanzol, Prodactif, Quellada, Sang-gamma, Scabecid, Scabene, Spritz-Rapidin, Spritzlindane, Spruehpflanzol, Streunex, TAP 85, Tri-6, Verindal Ultra, Viton, γ-1,2,3,4,5,6-Hexachlorocyclohexane, γ-666, γ-BHC, γ-Benzene hexachloride, γ-Benzohexachloride, γ-HCH, γ-Hexachloran, γ-Hexachlorane, γ-Hexachlorobenzene, γ-Hexachlorocyclohexane, γ-Lindane
IUPAC Name 1,2,3,4,5,6-hexachlorocyclohexane
SMILES Cl[C@@H]1[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@H](Cl)[C@H]1Cl

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5+,6+
Formula Weight 287.8601
Chemical Name or Material gamma-HCH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.