missing translation for 'onlineSavingsMsg'
Learn More

IKA™ Linea Pipette Rack

Product Code. 16258551 Shop All IKA Products
Change view
Click to view available options
For Use With (Equipment):
IKA™ Pette Vario Pipettes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16258551 IKA™ Pette Vario Pipettes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16258551 Supplier IKA™ Supplier No. 0020018987
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pipette rack designed to hold a variety of pipette sizes.

  • For use with pipettes ranging from 0.2 μL to 1000 μL
TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Equipment) IKA™ Pette Vario Pipettes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.