Learn More
Edulab Linear Air Track with accessories
Product Code. 30280263
Change view
Click to view available options
Length (English):
1.5 m
Unit Size:
1 set
Product Code. Length (English) Unit Size
30280263 1.5 m 1 set
1 options
Product Code. 30280263 Supplier Edulab Supplier No. VTT12305003
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
An affordable way to study dynamics in a friction-free setting, ideal for observing collisions and demonstrating conservation of momentum.
  • Allows experiments in velocity, force, acceleration, energy transfer, simple harmonic motion, and motion on both level and inclined planes
  • Constructed from durable extruded aluminum for strength and stability in laboratory use
  • Gliders included, designed to work with standard slotted masses
  • Comes with an accessory pack containing vehicles, mass attachments, catapults, and buffers
  • Supports the study of conservation of momentum and other fundamental physics principles
Specifications
For Use With (Equipment) Linear Air Track
Product Type Air Track
Quantity 1
Length (English) 1.5 m
NOTE: This air track should be used with the Edulab Linear Air Track Blower (VTT12303290).