Learn More
Checkers Linebacker™ 5 Channel Heavy Duty End Caps
Product Code. 17712428
Change view
Click to view available options
Color:
Orange
Unit Size:
Each
Product Code. Color Unit Size
17712428 Orange Each
1 options
Product Code. 17712428 Supplier Checkers Supplier No. PLBS0007OR
  • ONLINE EXCLUSIVE PRICE
    153.20 €
    123.95 € / Pack of 1
    Save 29.25 €
    19% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Linebacker™ 5-channel end caps for use with Linebacker™ heavy duty cable protector. Provides a beveled edge at the end of cable protectors to allow for a safer crossing for vehicles and pedestrian traffic while protecting valuable electrical cables and hose lines from damage.
  • Linebacker™ 5-channel end caps for use with Linebacker™ heavy duty cable protector
  • Provides a bevelled edge at the end of cable protectors to allow for a safer crossing for vehicles and pedestrian traffic while protecting valuable electrical cables and hose lines from damage
  • 5-channel end caps are sold in pairs (male and female)
Specifications
Dimensions (L x W x H) 51 x 19 x 6 cm
Color Orange
Material Polyethylene
Type 5 Channel End Caps