missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL Lining for Glass Tapers
Click to view available options
Standard Taper Joint No.:
10/19
12/21
7/16
Unit Size:
Pack of 10
Description
- Specially designed to prevent machined glass joints from binding.
- Conforms to DIN 12242.
- SUITABLE FOR FOODSTUFF.
- Available in several different dimensions up to 45/40 cone sizes.
- PTFEs (Polytetrafluoroethylene) are polymers with high molecular weight that are widely used owing to their optimum chemical, electrical and thermal properties; their friction coefficient is very low.
- Practically total chemical inertia with reagents and solvents.
- They can be used in the more corrosive environments and do not contaminate the more sensitive materials in ultra-pure or corrosive applications.
- Very high thermal stability.
- Use temperature from -200°C to +300°C; thermal degradation does not occur
before +400°C. - Exceptional Electrical Properties.
- Extremely high electrical resistance low dielectric constant and low dielectric loss factor
- Outstanding non-stick properties
- Self-lubricating and shockproof features, toughness and flexibility even at low temperatures
- Chemical applications: very high chemical resistance to acids, alkali and solvents is typical of the fluoride polymers; it is therefore used for components in the petrochemical and chemical industry alimentary: physiologically inert, it is approved by some Authorities for uses in contact with foods electrical: excellent dielectric, self-extinguishing and stability in bad weather features make it increasingly used in this sector mechanical: its low friction coefficient makes it usable for very low load applications.
- The magnetic stir bars shown on the following pages have internal magnets in Alnico V that gives them of high duration and power features. These magnets are encapsulated in first quality PTFE cases.
- The strict quality controls applied to these products ensure correct positioning of the magnetic core, the quality of the external surface, breakage resistance and constancy of the thickness of the external covering.
Specifications
Specifications
| Standard Taper Joint No. | 7/16 |
| Type | Sleeve |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction