missing translation for 'onlineSavingsMsg'
Learn More

Linoleic acid, 95%

Product Code. 11362117
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362117 5 g 5g
11372117 25 g 25g
11382117 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362117 Supplier Thermo Scientific Alfa Aesar Supplier No. L07949.06
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 60-33-3 | C18H32O2 | 280.452 g/mol

Unsaturated μ-6 fatty acid Linoleic acid is involved in the production of quick-drying oils which are used in oil paints and varnishes. It is also used in beauty products due to its beneficial properties on the skin such as moisture retention. It is a key starting material for the synthesis of linoleyl alcohol and octadecanoic acid. It is used as a precursor of arachidonic acid (AA) and various prostaglandins. It may be used to improve the delivery and efficacy of anti-cancer drugs and in cancer protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Unsaturated µ-6 fatty acid Linoleic acid is involved in the production of quick-drying oils which are used in oil paints and varnishes. It is also used in beauty products due to its beneficial properties on the skin such as moisture retention. It is a key starting material for the synthesis of linoleyl alcohol and octadecanoic acid. It is used as a precursor of arachidonic acid (AA) and various prostaglandins. It may be used to improve the delivery and efficacy of anti-cancer drugs and in cancer protection.

Solubility
Miscible with dimethyl sulfoxide, ethanol, ether, petroleum ether, dimethylformamide, acetone and benzene. Insoluble in water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 60-33-3
Molecular Formula C18H32O2
Molecular Weight (g/mol) 280.452
MDL Number MFCD00064241
InChI Key OYHQOLUKZRVURQ-HZJYTTRNSA-N
Synonym linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
PubChem CID 5280450
ChEBI CHEBI:17351
IUPAC Name (9Z,12Z)-octadeca-9,12-dienoic acid
SMILES CCCCCC=CCC=CCCCCCCCC(=O)O

Spezifikation

Melting Point -12°C
Density 0.902
Boiling Point 230°C (16 mmHg)
Flash Point >110°C (230°F)
Odor Weak, Characteristic
Linear Formula CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
Refractive Index 1.4699
Quantity 5 g
Beilstein 1727101
Sensitivity Air sensitive
Merck Index 14,5505
Solubility Information Miscible with dimethyl sulfoxide,ethanol,ether,petroleum ether,dimethylformamide,acetone and benzene. Insoluble in water.
Formula Weight 280.45
Percent Purity 95%
Chemical Name or Material Linoleic acid
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.