missing translation for 'onlineSavingsMsg'
Learn More

Linolenic Acid, TRC

Product Code. 30133878
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133878 5 g 5g
17917724 1 g 1g
17928784 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133878 Supplier Toronto Research Chemicals Supplier No. TRCL4676505G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 463-40-1
Molecular Formula C18 H30 O2
Molecular Weight (g/mol) 278.43
Synonym 9,12,15-Octadecatrienoic acid, (9Z,12Z,15Z)-, 9,12,15-Octadecatrienoic acid, (Z,Z,Z)-, Linolenic acid (8CI), (9Z,12Z,15Z)-9,12,15-Octadecatrienoic acid, (Z,Z,Z)-Octadeca-9,12,15-trienoic acid, (all-Z)-9,12,15-Octadecatrienoic acid, 18:3n-3, 9,12,15-all-cis-Octadecatrienoic acid, 9-cis,12-cis,15-cis-Octadecatrienoic acid, 9Z,12Z,15Z-Octadecatrienoic acid, all-cis-9,12,15-Octadecatrienoic acid, alpha-Linolenic acid, cis,cis,cis-9,12,15-Octadecatrienoic acid, cis-9,cis-12,cis-15-Octadecatrienoic acid, cis-δ9,12,15-Octadecatrienoic acid, α-Linolenic acid
IUPAC Name (9Z,12Z,15Z)-octadeca-9,12,15-trienoic acid
SMILES CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C18H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h3-4,6-7,9-10H,2,5,8,11-17H2,1H3,(H,19,20)/b4-3-,7-6-,10-9-
Formula Weight 278.2246
Percent Purity >95%
Chemical Name or Material cis alpha-Linolenic acid (ALA)
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.