missing translation for 'onlineSavingsMsg'
Learn More

Linoleyl Iodide, TRC

Product Code. 30477580
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477580 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477580 Supplier Toronto Research Chemicals Supplier No. TRCL60839010MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Linoleyl Iodide
CAS 60839-74-9
Molecular Formula C18H33I
Quantity 10 mg
Synonym (6Z,9Z)-18-Iodo-6,9-octadecadiene, (Z,Z)-18-Iodo-6,9-octadecadiene
InChI Formula InChI=1S/C18H33I/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h6-7,9-10H,2-5,8,11-18H2,1H3/b7-6-,10-9-
SMILES CCCCC\C=C/C\C=C/CCCCCCCCI
IUPAC Name (6Z,9Z)-18-iodooctadeca-6,9-diene
Molecular Weight (g/mol) 376.36
Formula Weight 376.1627
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.