missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ 2 Gel Transfer Device

Product Code. 15217995
Change view
Click to view available options
Quantity:
1 Device
Unit Size:
1 unit
Product Code. Quantity unitSize
15217995 1 Device 1 unit
1 options
This item is not returnable. View return policy

Product Code. 15217995

Brand: Invitrogen™ IB21001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Requires

iBlot 2 device and transfer stacks

Performs Western blotting transfer simply, efficiently, and reliably, within seven minutes and without the need for liquid buffers

The iBlot 2 Gel Transfer Device is an integral part of the iBlot 2 Dry Blotting System, which consists of the transfer device and consumable transfer stacks that contain the required buffers and transfer membrane (nitrocellulose or PVDF).

  • Complete protein transfer in 7 minutes or less
  • High detection sensitivity and even transfer
  • Increased blotting reliability and reproducibility
  • Flexible gel size formats and membrane types
  • A simple, user-friendly system
  • Options to create new custom programs
  • Built-in tutorial and application notes
  • High-quality transfer stacks that are more compact than before

Rapid, High-quality Protein Transfers

  • For dry electroblotting of proteins from mini-, midi-, and E-PAGE™ gels onto nitrocellulose or PVDF membranes for Western detection
  • Offers high-quality transfer, convenience, and speed, producing crisp, clear bands that remain sharp and straight, with exceptional transfer efficiency
  • Allows users to create custom programs
iBlot 2 Transfers Save Time
  • With the iBlot 2 system, there is no need to prepare buffers, pretreat the gel, or clean up after blotting
  • The total preparation and run time is normally less than 10 minutes per blot for significant time savings compared to conventional Western transfer techniques
How It Works
  • Buffer ion reservoirs are incorporated into the gel matrix (transfer stacks) instead of buffer tanks or soaked papers
  • The high density of ions in the gel matrix enables rapid protein transfer
  • During blotting, the copper anode does not generate oxygen gas as a result of water electrolysis, reducing blot distortion (conventional protein transfer techniques, including wet, semi-wet, and semi-dry, use inert electrodes that generate oxygen)
  • Transfer time is reduced by the shortened distance between electrodes, high field strength, and high current
  • Trapped air bubbles, often created during the manual preparation of the blotting sandwich layers, are easy to avoid due to the de-bubbling design that promotes even and complete transfer

Proteins, Expression, Isolation and Analysis, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Application) Western Blot
For Use With (Equipment) iBlot™
Gel Compatibility Bolt Bis-Tris Plus Gel, E-PAGE Gel, Novex Midi Gel, NuPAGE Gel, Novex Mini Gel
Membrane Compatibility Nitrocellulose, PVDF
Mode of Transfer Dry
Product Line iBlot
Running Time ∼7 min.
Throughput 1 Midi or 2 Mini Gels
Type Gel Transfer Device
Quantity 1 Device
Running Dimension Horizontal
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.