missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ MessengerMAX™ Transfection Reagent

Product Code. 15321734
Change view
Click to view available options
Quantity:
0.1 mL
0.3 mL
0.75 mL
1.5 mL
15 mL
Unit Size:
0.1mL
0.3mL
0.75mL
1.5mL
15mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15321734 0.1 mL 0.1mL
15397974 0.3 mL 0.3mL
15364584 0.75 mL 0.75mL
15361844 1.5 mL 1.5mL
15374584 15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15321734 Supplier Invitrogen™ Supplier No. LMRNA001
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Maximize mRNA transfection with Lipofectamine MessengerMAX Transfection Reagent. This optimized lipid-based mRNA transfection reagent enables efficient mRNA delivery with a simple and scalable protocol for a wide variety of cell lines.

Lipofectamine MessengerMAX mRNA Transfection Reagent delivers amazing mRNA transfection efficiency, enabling improved application outcomes and more biologically relevant research. That's because our novel lipid-based technology is optimized for efficient mRNA transfection without the nuclear entry step that is required with DNA. When combined with our mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG, superior mRNA expression is achievable in a wide range of cell types.

Successfully transfect more predictive cell models with a reagent that provides:

  • Rapid protein expression—enables quick translation of mRNA into protein, with no risk of genomic integration, accelerating experimental timelines
  • Broad cell type applicability—versatile use in a wide range of cell lines, including neurons and primary cell types
  • Minimal cytotoxicity—maintains cell viability and health, ideal for sensitive cells and long-term studies
  • High-throughput compatibility—suitable for high-throughput screening, facilitating large-scale mRNA transfection studies
  • Higher genome editing—up to 10X higher cleavage using CRISPR-Cas9 mRNA

Enable high transfection efficiency in novel genome editing applications

Lipofectamine MessengerMAX reagent helps increase the likelihood of successful cleavage and recombination through highly efficient Cas9 mRNA transfection, ultimately maximizing the efficiency of genetic modifications and simplifying the downstream processes.

Efficiently transfect difficult-to-transfect cells

Lipofectamine MessengerMAX reagent is designed for efficient mRNA transfection into neurons and a broad spectrum of difficult-to-transfect primary cells. This eliminates the need for electroporation and provides a >2-fold improvement in transfection efficiency compared to other lipid-based reagents.

Leverage advantages of mRNA transfection

mRNA transfection does not require nuclear uptake, leading to faster protein expression than DNA transfection, since mRNA translation occurs in the cytoplasm. Additionally, mRNA poses no risk of genomic integration and maintains high transfection efficiency regardless of the cell cycle stage.

Working in vivo? Try our lipid nanoparticle products Vivofectamine™ VF232 Liver LNP Composition in Ethanol (Cat. No. VF232LVCE) and Vivofectamine™ VF233 IM LNP Composition in Ethanol (Cat. No. VF233IMCE) for mRNA transfection in vivo.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Cell Type Established Cell Lines, Neuronal Cells, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type mRNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
Product Line Lipofectamine
Product Type Transfection Reagent
Quantity 0.1 mL
Serum Compatible Yes
Shipping Condition Room Temperature
Show More Show Less
Can I deliver single-stranded oligonucleotides with Lipofectamine MessengerMAX Transfection Reagent?

If you are delivering a single-stranded oligonucleotide on its own, we recommend using Lipofectamine RNAiMAX Transfection Reagent. However, Lipofectamine MessengerMAX Transfection Reagent can be used for genome editing purposes when Cas9 mRNA is co-delivered with a sgRNA and a single-stranded or double-stranded donor template.

How do I use Lipofectamine MessengerMAX Reagent to deliver both my GeneArt Cas9 mRNA and IVT gRNA together?

Lipofectamine MessengerMAX Reagent may be used for both single gRNA delivery or multiplexing (gRNA for multiple targets) purposes with high transfection efficiency when used with GeneArt Cas9 mRNA. For a detailed protocol, please refer to the GeneArt Cas9 mRNA manual (https://www.thermofisher.com/order/catalog/product/A25640?ICID=search-a25640). We recommend using either the Invitrogen GeneArt Genomic Cleavage Detection Kit (Cat. No. A24732) or Invitrogen GeneArt Genomic Cleavage Selection Kit (Cat. No. A27663) for mutant analysis.

Which cell types can be used with Lipofectamine MessengerMAX Transfection Reagent?

Lipofectamine MessengerMAX Reagent demonstrates low toxicity and high transfection efficiency for all cell types (easy or difficult-to-transfect, primary, and stem cells).

When would I use Lipofectamine MessengerMAX Reagent over other transfection reagents?

Lipofectamine MessengerMAX Regent is an excellent reagent for co-delivery of Invitrogen GeneArt CRISPR Cas9 mRNA with in vitro transcribed gRNA for geneome editing purposes. Lipofectamine MessengerMAX Reagent has the added benefit of flexibly delivering short dsDNA or HDR templates (0.5-1 kb) which can be ordered through the Invitrogen GeneArt Strings services.

What is the Invitrogen Lipofectamine MessengerMAX Transfection Reagent?

Lipofectamine MessengerMAX Transfection Reagent is an animal origin-free transfection reagent, especially formulated for the delivery of mRNA, small RNA (eg.,CRISPR IVT gRNA, siRNA, or miRNA), and short dsDNA or HDR templates (0.5-1 kb). Lipofectamine MessengerMAX Reagent is an excellent reagent choice for CRISPR-mediated genome editing applications.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.