missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ MessengerMAX™ Transfection Reagent

Lipofectamine™ MessengerMAX™ Transfection Reagent

Brand:  Invitrogen™ LMRNA015

1236.90 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.



Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15361844

  • 1302.00 € / 1.50mL
Estimated Shipment: 27-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Lipofectamine™ MessengerMAX mRNA Transfection Reagent delivers amazing transfection efficiency in neurons and a broad-spectrum of primary cells, enabling improved application outcomes and more biologically relevant research. That's because our novel lipid nanoparticle technology is optimized to deliver the highest amount of mRNA possible without the nuclear entry step that is required with DNA.

Successfully transfect more predictive cell models with a reagent that provides:
  • Amazing transfection efficiency in neurons and primary cell types
  • Faster protein expression with no risk of genomic integration
  • Up to 10x higher cleavage using mRNA CRISPRs

    Enable high transfection efficiency in novel genome editing applications
    Lipofectamine™ MessengerMAX™ reagent helps increase the likelihood of successful cleavage and recombination with GeneArt™ CRISPR Nuclease mRNA through highly efficient transfection, ultimately maximizing the efficiency of genetic modifications and simplifying the downstream processes.

    Efficiently transfect difficult-to-transfect cells
    Lipofectamine™ MessengerMAX™ reagent is designed to transfect a higher amount of mRNA into neurons and a broad spectrum of difficult-to-transfect primary cells. This eliminates the need for electroporation or viruses, and provides a > 2-fold improvement in transfection efficiency compared to other lipid based reagents.

    Leverage advantages of mRNA transfection
    Transfection of mRNA does not require nuclear uptake, which results in faster protein expression than DNA transfection since translation of mRNA occurs in the cytoplasm. Also, mRNA has no risk of genomic integration and transfection efficiency is cell cycle independent.
  • Specifications

    Specifications

    Store in refrigerator (2–8°C).
    6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
    Lipid-based Transfection
    Sustainable packaging
    Transfection Reagent
    Yes
    Established Cell Lines, Neuronal Cells, Stem Cells, Primary Cells, Hard-to-Transfect Cells
    mRNA
    Transfection
    Lipofectamine™
    1.5 mL
    Room Temperature
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.