missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ Liq-Glass Temp Electrode
Specifications
Specifications
| Accuracy (mV) | 0 ± 20mV |
| Accuracy (Temperature) | -10°C to +100°C |
| Diameter (Metric) | 12 mm |
| Electrical Requirements | 0 ± 20 mV, 57 to 59 mV |
| Length (Metric) | 120 mm |
| pH Range | 0 to 14 |
| Temperature (Metric) Range | -10°C to +100°C |
| Certifications/Compliance | Yes, with measured values in Buffer pH 4.01 and pH 7.00 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?