missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Liquid RNase A

Product Code. 15849087
Click to view available options
Quantity:
2 x 1.25 mL
Unit Size:
2.5mL
This item is not returnable. View return policy

Product Code. 15849087

Brand: Macherey-Nagel™ 740397

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Liquid Rnase A is easy to handle without dissolving. It is included in many NucleSpin and NucleoBond kits, where it is added directly to a resuspension buffer in order to prepare samples (e.g., pelleted bacteria) for cell lysis.

  • The Liquid RNase A is primarily intended to be used with NucleoSpin™ and NucleoBond™ kits
  • Has the same specifications as the enzyme provided with all MACHEREY-NAGEL products.
  • Used in the suspension of samples for the purpose of cell lysis
  • Can be used to digest nucleic acids enzymatically when eluted RNA/DNA fractions contain a fraction of the other nucleic acid

Specifications

Concentration 100 mg/mL
Color Colorless
Components Rnase liquid (2.5 - <10%), di-Sodium hydrogencitrate (<10%), Water
pH 4.5 to 5.5
For Use With (Equipment) NucleoSpin and NucleoBond kits
CAS 9001-99-4L
Content And Storage Room temperature (18°C to 25°C)
Source Bovine pancreas
Quantity 2 x 1.25 mL
Product Type Rnase A Enzyme Solution
Form Ready-to-Use Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.