Learn More
Grant Instruments™ Liquid Sanitizer
This liquid sanitizer is for use in unstirred water baths to prevent bacterial and fungal growth.
Brand: Grant Instruments™ SANIT1
241.45 EUR valid until 2024-05-31
Use promo code "21679" to get your promotional price.
Additional Details : CAS Number : 7722-84-1 Weight : 4.90000kg
Description
- Cleaning solution for laboratory water baths, static water systems and flotation baths
- Ready-to-use solution and comes with a measuring cup for simple and accurate manual dosing
- Just add 10 mL of solution for every 1 L of fresh water in a clean water bath/system
- Re-dose weekly
- After 4 weeks, completely empty water from bath/system, refill with fresh water and repeat dosing procedure
- Non-corrosive to stainless steel and other materials associated with water baths/systems when used at recommended dosage levels
Specifications
Solution | |
Liquid | |
Grant™ | |
Sterile | |
Clear | |
Single-use | |
No | |
Plastic Bottle | |
Water Soluble | |
1 L |
No | |
<5 | |
Unscented | |
Not Autoclavable | |
1.028 kg/cm3 at 15°C | |
Non-filtered | |
Grant™ JBA, JBN, SAP Baths, SBB Boiling Baths, LSB, OLS Shaking Baths, Stirred Baths, Ultrasonic Baths | |
4 x 1 L Bottles | |
1.005 cP at 20°C |
Safety and Handling
- Liquid sanitiser
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P235-Keep cool.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P403-Store in a well-ventilated place.
- P405-Store locked up.
- P410-Protect from sunlight.
Supplemental information
- MIXTURE LIST-Contains : hydrogen peroxide