missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lissamine rhodamine B sulfonyl chloride, pure

Product Code. 10167940 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10167940

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62796-29-6 | C27H29ClN2O6S2 | 577.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62796-29-6
Molecular Formula C27H29ClN2O6S2
Molecular Weight (g/mol) 577.107
MDL Number MFCD00042001
InChI Key YERWMQJEYUIJBO-UHFFFAOYSA-N
Synonym Sulforhodamine B sulfonyl chloride
PubChem CID 65223
IUPAC Name 5-chlorosulfonyl-2-[3-(diethylamino)-6-diethylazaniumylidenexanthen-9-yl]benzenesulfonate
SMILES CCN(CC)C1=CC2=C(C=C1)C(=C3C=CC(=[N+](CC)CC)C=C3O2)C4=C(C=C(C=C4)S(=O)(=O)Cl)S(=O)(=O)[O-]

Specifications

Assay Percent Range 99+%
Infrared Spectrum Authentic
Formula Weight 577.11
Grade Pure
Packaging Glass bottle
UV Spectrometry Molar ext.%gt; 85, 000L/mol.cm(lambda 564 +/- 3nm) in dichloromethane
Color Green
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Lissamine rhodamine B sulfonyl chloride, 99+%
compliance-icons
Product Identifier
  • Lissamine rhodamine B sulfonyl chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.