missing translation for 'onlineSavingsMsg'
Learn More

Lisuride Maleate, TRC

Product Code. 30011215
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011215 10 mg 10mg
30006975 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011215 Supplier Toronto Research Chemicals Supplier No. TRCL46907810MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19875-60-6
Molecular Formula C20H26N4O •C4H4O4
Molecular Weight (g/mol) 338.4511607
Synonym 3-(9,10-Didehydro-6-methylergolin-8α-yl)-1,1-diethyl-urea Maleate, N'-[(8α)-9,10-Didehydro-6-methylergolin-8-yl]-N,N-diethyl-urea (Z) 2-Butenedioate, Ergoline Urea Deriv., Indolo[4,3-fg]quinoline Urea Deriv., Apodel, Cuvalit, Dopergin, Eunal, Lisuride Hydrogen Maleate, Lysenyl, Lysenyl Bimaleate, Lysenyl Hydrogen Maleate, Revanil;
SMILES OC(/C=C\C(O)=O)=O.CN1[C@](C2=C[C@H](NC(N(CC)CC)=O)C1)([H])CC3=CNC4=C3C2=CC=C4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H26N4O.C4H4O4/c1-4-24(5-2)20(25)22-14-10-16-15-7-6-8-17-19(15)13(11-21-17)9-18(16)23(3)12-14;5-3(6)1-2-4(7)8/h6-8,10-11,14,18,21H,4-5,9,12H2,1-3H3,(H,22,25);1-2H,(H,5,6)(H,7,8)/b;2-1-/t14-,18+;/m0./s1
Chemical Name or Material Lisuride Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.