missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetate dihydrate, Reagent Grade, Honeywell

Product Code. p-7161448 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15667910 250 g 250g
15657910 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15667910 Supplier Honeywell Research Chemicals Supplier No. L6883250G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6108-17-4
Molecular Formula C2H7LiO4
Molecular Weight (g/mol) 102.01
MDL Number MFCD00066949
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
Synonym Acetic acid lithium salt
IUPAC Name lithium(1+) acetate dihydrate
SMILES [Li+].O.O.CC([O-])=O

Specifications

CAS Max % ≤100.00
Physical Form Solid
Quantity 250 g
Beilstein 3564320
Grade Reagent
Packaging Plastic Bottle
Density 1.3 g/cm3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.